Assay Details
| Gene Symbol: SGPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005686103.3 | XP_005686160.2 | 2 - 3 | 977 | 150 | |
| GenBank mRNA | KA304631.1 | - | 2 - 3 | 1018 | 150 | |
More Information
Set Membership: |
|

SGPP1
Genomic Map