Assay Details
| Gene Symbol: SP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285760.1 | NP_001272689.1 | 2 - 3 | 1681 | 60 | |
| XM_018047544.1 | XP_017903033.1 | 3 - 4 | 2118 | 60 | ||
| XM_018047545.1 | XP_017903034.1 | 3 - 4 | 1625 | 60 | ||
| GenBank mRNA | HM236311.1 | - | 2 - 3 | 1681 | 60 | |
More Information
Set Membership: |
|

SP1
Genomic Map