Assay Details
| Gene Symbol: MINDY4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013963359.2 | XP_013818813.2 | 14 - 15 | 1912 | 57 | |
| XM_018047112.1 | XP_017902601.1 | 15 - 16 | 1911 | 57 | ||
| GenBank mRNA | JT640591.1 | - | 9 - 10 | 1249 | 57 | |
More Information
Set Membership: |
|

MINDY4
Genomic Map