Assay Details
| Gene Symbol: C16H1orf21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060709.1 | XP_017916198.1 | 4 - 5 | 527 | 133 | |
| XM_018060710.1 | XP_017916199.1 | 4 - 5 | 747 | 133 | ||
| GenBank mRNA | JO392940.2 | - | 4 - 5 | 679 | 133 | |
More Information
Set Membership: |
|

C16H1orf21
Genomic Map