Assay Details
| Gene Symbol: LOC102180747 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013964712.2 | XP_013820166.2 | 1 - 2 | 443 | 80 | |
| XM_018049561.1 | XP_017905050.1 | 1 - 2 | 444 | 80 | ||
More Information
Set Membership: |
|

LOC102180747
Genomic Map