Assay Details
| Gene Symbol: LRCH4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040376.1 | XP_017895865.1 | 2 - 3 | 579 | 55 | |
| XM_018040377.1 | XP_017895866.1 | 2 - 3 | 578 | 55 | ||
| GenBank mRNA | JO401530.2 | - | 11 - 12 | 1084 | 55 | |
| JO438453.2 | - | 8 - 9 | 802 | 55 | ||
More Information
Set Membership: |
|

LRCH4
Genomic Map