Assay Details
| Gene Symbol: UBE2M | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063555.1 | XP_017919044.1 | 2 - 3 | 766 | 65 | |
| GenBank mRNA | JO633835.2 | - | 2 - 3 | 742 | 65 | |
| KA325985.1 | - | 4 - 5 | 733 | 65 | ||
More Information
Set Membership: |
|

UBE2M
Genomic Map