Assay Details
| Gene Symbol: KLF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056510.1 | XP_017911999.1 | 2 - 3 | 1236 | 87 | |
| GenBank mRNA | KA306136.1 | - | 2 - 3 | 911 | 87 | |
| KU041751.1 | - | 1 - 2 | 863 | 87 | ||
More Information
Set Membership: |
|

KLF5
Genomic Map