Assay Details
| Gene Symbol: ZFAND5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052069.1 | XP_017907558.1 | 4 - 5 | 1281 | 77 | |
| XM_018052070.1 | XP_017907559.1 | 4 - 5 | 503 | 77 | ||
| GenBank mRNA | JO428804.2 | - | 2 - 3 | 1827 | 77 | |
More Information
Set Membership: |
|

ZFAND5
Genomic Map