Assay Details
| Gene Symbol: RNPS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018040423.1 | XP_017895912.1 | 6 - 7 | 892 | 61 | |
| XM_018040424.1 | XP_017895913.1 | 6 - 7 | 1259 | 61 | ||
| XM_018040425.1 | XP_017895914.1 | 6 - 7 | 1207 | 61 | ||
| XM_018040426.1 | XP_017895915.1 | 5 - 6 | 694 | 61 | ||
| GenBank mRNA | JO434638.2 | - | 3 - 4 | 415 | 61 | |
| JO643468.2 | - | 6 - 7 | 863 | 61 | ||
More Information
Set Membership: |
|

RNPS1
Genomic Map