Assay Details
| Gene Symbol: BMPR1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285575.1 | NP_001272504.1 | 7 - 8 | 850 | 65 | |
| XM_013964506.2 | XP_013819960.1 | 7 - 8 | 1512 | 65 | ||
| XM_013964507.2 | XP_013819961.1 | 7 - 8 | 1226 | 65 | ||
| XM_013964508.2 | XP_013819962.1 | 7 - 8 | 3154 | 65 | ||
| XM_013964509.2 | XP_013819963.1 | 7 - 8 | 3149 | 65 | ||
| XM_018049148.1 | XP_017904637.1 | 9 - 10 | 1058 | 65 | ||
| XM_018049150.1 | XP_017904639.1 | 10 - 11 | 1076 | 65 | ||
| XM_018049151.1 | XP_017904640.1 | 7 - 8 | 1051 | 65 | ||
| XM_018049152.1 | XP_017904641.1 | 7 - 8 | 1251 | 65 | ||
| XM_018049153.1 | XP_017904642.1 | 9 - 10 | 1053 | 65 | ||
| XM_018049154.1 | XP_017904643.1 | 7 - 8 | 872 | 65 | ||
| GenBank mRNA | DQ005917.1 | - | 3 - 4 | 359 | 65 | |
| DQ060443.1 | - | 7 - 8 | 850 | 65 | ||
| DQ666418.1 | - | 6 - 7 | 790 | 65 | ||
| EU847285.1 | - | 6 - 7 | 781 | 65 | ||
| EU847290.1 | - | 6 - 7 | 781 | 65 | ||
| FJ712710.1 | - | 6 - 7 | 804 | 65 | ||
| GU224270.1 | - | 1 - 2 | 119 | 65 | ||
| GU359045.1 | - | 6 - 7 | 781 | 65 | ||
| JN100107.1 | - | 6 - 7 | 788 | 65 | ||
| KF758765.1 | - | 8 - 9 | 937 | 65 | ||
More Information
Set Membership: |
|

BMPR1B
Genomic Map