Assay Details
| Gene Symbol: IGFBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285678.1 | NP_001272607.1 | 2 - 3 | 506 | 60 | |
| GenBank mRNA | EU559624.1 | - | 2 - 3 | 506 | 60 | |
| JO425166.1 | - | 2 - 3 | 830 | 60 | ||
| JO559459.2 | - | 2 - 3 | 777 | 60 | ||
| JQ341162.1 | - | 2 - 3 | 653 | 60 | ||
More Information
Set Membership: |
|

IGFBP4
Genomic Map