Assay Details
| Gene Symbol: FKBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048639.1 | XP_017904128.1 | 7 - 8 | 1091 | 65 | |
| GenBank mRNA | JO497445.2 | - | 3 - 4 | 1243 | 65 | |
More Information
Set Membership: |
|

FKBP4
Genomic Map