Assay Details
| Gene Symbol: SZRD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054694.1 | XP_017910183.1 | 3 - 4 | 424 | 61 | |
| XM_018054695.1 | XP_017910184.1 | 3 - 4 | 421 | 61 | ||
| GenBank mRNA | JO442204.2 | - | 3 - 4 | 407 | 61 | |
More Information
Set Membership: |
|

SZRD1
Genomic Map