Assay Details
| Gene Symbol: CHAF1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052761.1 | XP_017908250.1 | 11 - 12 | 1240 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CHAF1B| Gene Symbol: CHAF1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018052761.1 | XP_017908250.1 | 11 - 12 | 1240 | 69 | |
Set Membership: |
|