Assay Details
| Gene Symbol: LOC102171874 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005684175.3 | XP_005684232.3 | 17 - 18 | 3224 | 86 | |
| GenBank mRNA | JO424743.1 | - | 17 - 18 | 3128 | 86 | |
More Information
Set Membership: |
|

LOC102171874
Genomic Map