Assay Details
| Gene Symbol: SEPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697703.3 | XP_005697760.1 | 3 - 4 | 366 | 60 | |
| GenBank mRNA | JO329971.2 | - | 8 - 9 | 1213 | 60 | |
More Information
Set Membership: |
|

SEPT1
Genomic Map