Assay Details
| Gene Symbol: MAPK12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018048956.1 | XP_017904445.1 | 8 - 9 | 1365 | 67 | |
| XM_018048957.1 | XP_017904446.1 | 10 - 11 | 1400 | 67 | ||
| XM_018048959.1 | XP_017904448.1 | 9 - 10 | 2654 | 67 | ||
| GenBank mRNA | JO586978.2 | - | 7 - 8 | 536 | 67 | |
More Information
Set Membership: |
|

MAPK12
Genomic Map