Assay Details
| Gene Symbol: NFS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688676.3 | XP_005688733.2 | 3 - 4 | 403 | 58 | |
| GenBank mRNA | JO430734.2 | - | 10 - 11 | 1649 | 58 | |
More Information
Set Membership: |
|

NFS1
Genomic Map