Assay Details
| Gene Symbol: EXTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018056208.1 | XP_017911697.1 | 9 - 10 | 2402 | 61 | |
| XM_018056218.1 | XP_017911707.1 | 8 - 9 | 2348 | 61 | ||
| XM_018056228.1 | XP_017911717.1 | 8 - 9 | 2294 | 61 | ||
| XM_018056237.1 | XP_017911726.1 | 7 - 8 | 2240 | 61 | ||
| XM_018056244.1 | XP_017911733.1 | 9 - 10 | 2402 | 61 | ||
| GenBank mRNA | JO420045.2 | - | 3 - 4 | 883 | 61 | |
More Information
Set Membership: |
|

EXTL1
Genomic Map