Assay Details
| Gene Symbol: SRP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685480.3 | XP_005685537.1 | 4 - 5 | 342 | 71 | |
| GenBank mRNA | JO579028.1 | - | 1 - 2 | 751 | 71 | |
More Information
Set Membership: |
|

SRP14
Genomic Map