Assay Details
| Gene Symbol: NAAA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018049595.1 | XP_017905084.1 | 2 - 3 | 495 | 61 | |
| GenBank mRNA | JO580264.1 | - | 2 - 3 | 506 | 61 | |
| JT762435.1 | - | 1494 | 61 | |||
More Information
Set Membership: |
|

NAAA
Genomic Map