Assay Details
| Gene Symbol: CSNK1G2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018050405.1 | XP_017905894.1 | 8 - 9 | 1322 | 61 | |
| GenBank mRNA | JO621695.2 | - | 8 - 9 | 1366 | 61 | |
More Information
Set Membership: |
|

CSNK1G2
Genomic Map