Assay Details
| Gene Symbol: LOC102184120 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018046836.1 | XP_017902325.1 | 5 - 6 | 2006 | 75 | |
| XM_018046837.1 | XP_017902326.1 | 5 - 6 | 2005 | 75 | ||
| XM_018046838.1 | XP_017902327.1 | 5 - 6 | 2006 | 75 | ||
More Information
Set Membership: |
|

LOC102184120
Genomic Map