Assay Details
| Gene Symbol: RPAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005680040.3 | XP_005680097.1 | 9 - 10 | 1324 | 72 | |
| XM_018047885.1 | XP_017903374.1 | 9 - 10 | 1114 | 72 | ||
| XM_018047886.1 | XP_017903375.1 | 9 - 10 | 1085 | 72 | ||
More Information
Set Membership: |
|

RPAP3
Genomic Map