Assay Details
| Gene Symbol: SPAG8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683853.3 | XP_005683910.1 | 2 - 3 | 1573 | 69 | |
| XM_018052153.1 | XP_017907642.1 | 2 - 3 | 1573 | 69 | ||
| XM_018052154.1 | XP_017907643.1 | 2 - 3 | 1573 | 69 | ||
More Information
Set Membership: |
|

SPAG8
Genomic Map