Assay Details
| Gene Symbol: MEF2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051481.1 | XP_017906970.1 | 2 - 3 | 293 | 67 | |
| GenBank mRNA | JN967622.1 | - | 2 - 3 | 299 | 67 | |
| JO584953.2 | - | 6 - 7 | 1078 | 67 | ||
More Information
Set Membership: |
|

MEF2B
Genomic Map