Assay Details
| Gene Symbol: TWISTNB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005679024.3 | XP_005679081.2 | 1 - 2 | 274 | 103 | |
| GenBank mRNA | KA308609.1 | - | 1 - 2 | 280 | 103 | |
More Information
Set Membership: |
|

TWISTNB
Genomic Map