Assay Details
| Gene Symbol: KIF3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013968734.2 | XP_013824188.2 | 8 - 9 | 2246 | 55 | |
| XM_013968735.2 | XP_013824189.2 | 7 - 8 | 2122 | 55 | ||
| XM_018057697.1 | XP_017913186.1 | 8 - 9 | 2246 | 55 | ||
| GenBank mRNA | KA342375.1 | - | 2 - 3 | 3604 | 55 | |
More Information
Set Membership: |
|

KIF3B
Genomic Map