Assay Details
| Gene Symbol: DSE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018053037.1 | XP_017908526.1 | 4 - 5 | 1066 | 80 | |
| XM_018053038.1 | XP_017908527.1 | 4 - 5 | 1066 | 80 | ||
| XM_018053039.1 | XP_017908528.1 | 3 - 4 | 578 | 80 | ||
| XM_018053040.1 | XP_017908529.1 | 3 - 4 | 643 | 80 | ||
More Information
Set Membership: |
|

DSE
Genomic Map