Assay Details
| Gene Symbol: RNF167 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064528.1 | XP_017920017.1 | 8 - 9 | 1435 | 65 | |
| XM_018064530.1 | XP_017920019.1 | 7 - 8 | 1259 | 65 | ||
| GenBank mRNA | KA348301.1 | - | 8 - 9 | 1373 | 65 | |
More Information
Set Membership: |
|

RNF167
Genomic Map