Assay Details
| Gene Symbol: LYPLAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005690576.3 | XP_005690633.2 | 1 - 2 | 130 | 63 | |
| GenBank mRNA | JO447262.2 | - | 4 - 5 | 665 | 63 | |
More Information
Set Membership: |
|

LYPLAL1
Genomic Map