Assay Details
| Gene Symbol: MAPK7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018064745.1 | XP_017920234.1 | 3 - 4 | 1247 | 65 | |
| XM_018064746.1 | XP_017920235.1 | 2 - 3 | 236 | 65 | ||
| GenBank mRNA | JO530299.2 | - | 2 - 3 | 428 | 65 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018064746.1 transcripts.
Set Membership: |
|

MAPK7
Genomic Map