Assay Details
| Gene Symbol: EEF1AKNMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060313.1 | XP_017915802.1 | 2 - 3 | 1249 | 74 | |
| GenBank mRNA | JO443068.2 | - | 1 - 2 | 69 | 74 | |
More Information
Set Membership: |
|

EEF1AKNMT
Genomic Map