Assay Details
| Gene Symbol: LOC102180829 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043118.1 | XP_017898607.1 | 18 - 19 | 1937 | 68 | |
| XM_018043119.1 | XP_017898608.1 | 18 - 19 | 1934 | 68 | ||
| XM_018043120.1 | XP_017898609.1 | 18 - 19 | 1937 | 68 | ||
| XM_018043121.1 | XP_017898610.1 | 16 - 17 | 1585 | 68 | ||
| XM_018043123.1 | XP_017898612.1 | 17 - 18 | 1752 | 68 | ||
| XM_018043124.1 | XP_017898613.1 | 17 - 18 | 1643 | 68 | ||
| XM_018043125.1 | XP_017898614.1 | 17 - 18 | 1747 | 68 | ||
| XM_018043126.1 | XP_017898615.1 | 11 - 12 | 1063 | 68 | ||
| XM_018043127.1 | XP_017898616.1 | 11 - 12 | 1002 | 68 | ||
| XM_018043128.1 | XP_017898617.1 | 10 - 11 | 1103 | 68 | ||
| XM_018043129.1 | XP_017898618.1 | 10 - 11 | 934 | 68 | ||
| XM_018043130.1 | XP_017898619.1 | 4 - 5 | 666 | 68 | ||
| XM_018043131.1 | XP_017898620.1 | 11 - 12 | 961 | 68 | ||
| XM_018043133.1 | XP_017898622.1 | 4 - 5 | 475 | 68 | ||
| GenBank mRNA | JO520363.2 | - | 3 - 4 | 317 | 68 | |
More Information
Set Membership: |
|

LOC102180829
Genomic Map