Assay Details
| Gene Symbol: PPM1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675377.3 | XP_005675434.1 | 3 - 4 | 1509 | 65 | |
| GenBank mRNA | JT738072.1 | - | 5 - 6 | 1594 | 65 | |
| KA345866.1 | - | 3 - 4 | 799 | 65 | ||
More Information
Set Membership: |
|

PPM1L
Genomic Map