Assay Details
| Gene Symbol: UBE2W | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005689071.3 | XP_005689128.2 | 2 - 3 | 196 | 83 | |
| GenBank mRNA | JO605860.2 | - | 4 - 5 | 1304 | 83 | |
| KA324940.1 | - | 2 - 3 | 208 | 83 | ||
More Information
Set Membership: |
|

UBE2W
Genomic Map