Assay Details
| Gene Symbol: TAF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018039056.1 | XP_017894545.1 | 2 - 3 | 228 | 80 | |
| XM_018039057.1 | XP_017894546.1 | 2 - 3 | 228 | 80 | ||
| XM_018039058.1 | XP_017894547.1 | 2 - 3 | 228 | 80 | ||
More Information
Set Membership: |
|

TAF8
Genomic Map