Assay Details
Gene Symbol: RAB27A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018054226.1 | XP_017909715.1 | 6 - 7 | 793 | 61 | |
XM_018054227.1 | XP_017909716.1 | 7 - 8 | 869 | 61 | ||
XM_018054228.1 | XP_017909717.1 | 5 - 6 | 676 | 61 | ||
XM_018054229.1 | XP_017909718.1 | 5 - 6 | 596 | 61 | ||
XM_018054230.1 | XP_017909719.1 | 5 - 6 | 568 | 61 | ||
XM_018054231.1 | XP_017909720.1 | 5 - 6 | 574 | 61 | ||
GenBank mRNA | JO434641.2 | - | 1 - 2 | 701 | 61 | |
JO435913.2 | - | 1 - 2 | 701 | 61 | ||
JO612408.2 | - | 1 - 2 | 1493 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
