Assay Details
| Gene Symbol: FRMD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005685646.3 | XP_005685703.1 | 12 - 13 | 1296 | 67 | |
| XM_005685649.3 | XP_005685706.1 | 11 - 12 | 1234 | 67 | ||
| XM_018054345.1 | XP_017909834.1 | 9 - 10 | 1112 | 67 | ||
| XM_018054346.1 | XP_017909835.1 | 13 - 14 | 1398 | 67 | ||
| XM_018054347.1 | XP_017909836.1 | 12 - 13 | 1467 | 67 | ||
| XM_018054348.1 | XP_017909837.1 | 12 - 13 | 1336 | 67 | ||
| XM_018054349.1 | XP_017909838.1 | 9 - 10 | 1109 | 67 | ||
| XM_018054350.1 | XP_017909839.1 | 9 - 10 | 1102 | 67 | ||
| GenBank mRNA | KA344552.1 | - | 9 - 10 | 1087 | 67 | |
More Information
Set Membership: |
|

FRMD6
Genomic Map