Assay Details
| Gene Symbol: AHSA2P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055395.1 | XP_017910884.1 | 3 - 4 | 489 | 101 | |
| XM_018055396.1 | XP_017910885.1 | 3 - 4 | 489 | 101 | ||
| XM_018055397.1 | XP_017910886.1 | 3 - 4 | 489 | 101 | ||
| XM_018055398.1 | XP_017910887.1 | 3 - 4 | 259 | 101 | ||
| XM_018055399.1 | XP_017910888.1 | 3 - 4 | 267 | 101 | ||
More Information
Set Membership: |
|

AHSA2P
Genomic Map