Assay Details
| Gene Symbol: AHSA2P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018055395.1 | XP_017910884.1 | 6 - 7 | 812 | 92 | |
| XM_018055396.1 | XP_017910885.1 | 6 - 7 | 806 | 92 | ||
| XM_018055398.1 | XP_017910887.1 | 6 - 7 | 582 | 92 | ||
| XM_018055399.1 | XP_017910888.1 | 6 - 7 | 590 | 92 | ||
| XM_018055400.1 | XP_017910889.1 | 3 - 4 | 272 | 92 | ||
| XM_018055401.1 | XP_017910890.1 | 4 - 5 | 314 | 92 | ||
More Information
Set Membership: |
|

AHSA2P
Genomic Map