Assay Details
| Gene Symbol: THAP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018063377.1 | XP_017918866.1 | 2 - 3 | 470 | 63 | |
| XM_018063378.1 | XP_017918867.1 | 2 - 3 | 467 | 63 | ||
More Information
Set Membership: |
|

THAP8
Genomic Map