Assay Details
| Gene Symbol: ZZZ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013962713.2 | XP_013818167.1 | 4 - 5 | 526 | 60 | |
| XM_018045604.1 | XP_017901093.1 | 8 - 9 | 2509 | 60 | ||
| XM_018045608.1 | XP_017901097.1 | 8 - 9 | 2505 | 60 | ||
| XM_018045612.1 | XP_017901101.1 | 7 - 8 | 2430 | 60 | ||
| XM_018045617.1 | XP_017901106.1 | 9 - 10 | 2634 | 60 | ||
| XM_018045621.1 | XP_017901110.1 | 8 - 9 | 2506 | 60 | ||
| XM_018045625.1 | XP_017901114.1 | 7 - 8 | 2427 | 60 | ||
| GenBank mRNA | JO425977.1 | - | 7 - 8 | 1618 | 60 | |
More Information
Set Membership: |
|

ZZZ3
Genomic Map