Assay Details
| Gene Symbol: DHCR24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005678362.3 | XP_005678419.1 | 8 - 9 | 1536 | 62 | |
| GenBank mRNA | JO633377.2 | - | 8 - 9 | 1551 | 62 | |
| KA320690.1 | - | 8 - 9 | 1553 | 62 | ||
More Information
Set Membership: |
|

DHCR24
Genomic Map