Assay Details
| Gene Symbol: AP1S3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018058906.1 | XP_017914395.1 | 2 - 3 | 381 | 92 | |
| XM_018058907.1 | XP_017914396.1 | 2 - 3 | 386 | 92 | ||
| GenBank mRNA | KA350407.1 | - | 2 - 3 | 428 | 92 | |
More Information
Set Membership: |
|

AP1S3
Genomic Map