Assay Details
| Gene Symbol: HHEX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018041729.1 | XP_017897218.1 | 2 - 3 | 600 | 63 | |
| GenBank mRNA | JR130876.1 | - | 6 - 7 | 1281 | 63 | |
More Information
Set Membership: |
|

HHEX
Genomic Map