Assay Details
| Gene Symbol: ANAPC11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065455.1 | XP_017920944.1 | 2 - 3 | 345 | 97 | |
| XM_018065456.1 | XP_017920945.1 | 2 - 3 | 387 | 97 | ||
| GenBank mRNA | JO538640.2 | - | 2 - 3 | 243 | 97 | |
More Information
Set Membership: |
|

ANAPC11
Genomic Map