Assay Details
| Gene Symbol: MASTL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005687861.3 | XP_005687918.1 | 7 - 8 | 1076 | 61 | |
| XM_005687862.3 | XP_005687919.1 | 7 - 8 | 1076 | 61 | ||
| XM_018057021.1 | XP_017912510.1 | 7 - 8 | 1076 | 61 | ||
| XM_018057022.1 | XP_017912511.1 | 7 - 8 | 1076 | 61 | ||
| GenBank mRNA | JO590670.2 | - | 1 - 2 | 156 | 61 | |
More Information
Set Membership: |
|

MASTL
Genomic Map