Assay Details
| Gene Symbol: NAPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018062678.1 | XP_017918167.1 | 7 - 8 | 843 | 63 | |
| GenBank mRNA | JO413538.2 | - | 7 - 8 | 733 | 63 | |
| JO432635.2 | - | 7 - 8 | 713 | 63 | ||
More Information
Set Membership: |
|

NAPA
Genomic Map